Learn More
Mitutoyo™ Micrometro digitale IP54
Codice prodotto. 11737013
Encompass_Preferred
Cambia vista
Click to view available options
Range:
0 - 25 mm
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Range unitSize
11737013 0 - 25 mm Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11737013 Fornitore Mitutoyo™ N. del fornitore 293240
Aggiungi al carrello
Aggiungi al carrello
Highly robust digital micrometer with outstanding accuracy. Mitutoyo™ IP54 Digital Micrometer features extraordinary battery life.
  • Outstanding accuracy.
  • Oil-resistant material used for all plastic parts.
  • Extraordinary battery life of 2,4 years.
  • Excellent resistance against water and dust (IP65 protection level) enables this micrometer to be used in machining situations that include splashing coolant fluid.
Specifica
Product Type Micrometer
Display Digitale
Range 0 - 25 mm