Learn More
Iron(III) trifluoromethanesulfonate, TRC
Product Code. 30406627
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30406627 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30406627 Supplier Toronto Research Chemicals Supplier No. TRCI1661252G
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Iron(III) trifluoromethanesulfonate
CAS 63295-48-7
Molecular Formula C3F9FeO9S3
Quantity 2 g
Synonym Iron(Iii)Trifluoromethanesulfonate
InChI Formula InChI=1S/3CHF3O3S.Fe/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3
SMILES [Fe+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
IUPAC Name iron(3+);trifluoromethanesulfonate
Molecular Weight (g/mol) 503.05
Formula Weight 502.791