Learn More
Iron Solution for ICP, 1g/L, HNO3 Matrix, Fisher Chemical™
BIOPHARMA
Product Code. 12996184 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12996184 250 mL 250mL
12986184 100 mL 100mL
12906194 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12996184 Supplier Fisher Chemical Supplier No. S 119 250
  • ONLINE EXCLUSIVE PRICE
    108.00€
    78.86€ / 250mL
    Risparmia 29.14€
    26% Sconto
 Restricted Item
Spedizione stimata: 29-12-2026
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Packaging Glass Bottle
Analyte or Component Names Fe
Concentration 1.00±0.2 g/L
Element Fe
Solvent or Matrix Nitric Acid
Quantity 250 mL
compliance-icons
Product Identifier
  • ICP Standard Solution Iron (Fe) 1000mg/l in HNO
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands, forearms andface thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: iron trinitrate, nitric acid