Learn More
Chemsplash IRRADIATED Cape Hood
Product Code. 30058296
Change view
Click to view available options
Cleanroom Class:
Class 100-1000 (ISO 5-6)
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cleanroom Class Unit Size
30058296 Class 100-1000 (ISO 5-6) Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30058296 Supplier Chemsplash Supplier No. 5775.400.999
  • ONLINE EXCLUSIVE PRICE
    1035.18€
    876.44€ / pezzi 200
    Risparmia 158.74€
    15% Sconto
Spedizione stimata: 23-07-2027
Cape Hood is made from Chemsplash soft and microporous fabric. It is elasticated at the face and covers the hair, neck and top of the shoulders.
Chemsplash Cape Hood
  • Material: 55GSM Type PB 6B Fabric
  • Sterile Irradiated and Double Bagged
  • Elasticated Facial Area
  • Cape Style Fit
  • Silicone and Latex Free
  • Anti-Static Material
  • Non-Linting
  • PFAS Free
  • Conforms to ISO 14644-1:2015 Cleanroom 5 - 9
Description: The Chemsplash Cape Hood is made from Chemsplash's soft and microporous fabric, which is strong and resistant to chemical splash and dust. This cape hood is elasticated at the face and covers the hair, neck, and top of the shoulders. It is anti-static to meet EN1149-5:2008, lint-free, and suitable for cleanroom applications. Conforms to ISO 14644-1:2015 Cleanroom 5 - 9. Chemsplash accessories can be used to protect the wearer from the environment or the environment from the wearer, ensuring maximum comfort and wearability.
Specifications
Product Type Hood
Size One Size
Material Microporous
Color White
Closure Type Ties
Disposable Single-use
Latex Free Yes
Sterility Sterile
Cleanroom Class Class 100-1000 (ISO 5-6)