Learn More
ISE Sodium NA1000 PPM, Reagecon™
Product Code. 11779075 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11779075 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11779075 Supplier Reagecon Supplier No. ISENA5
  • ONLINE EXCLUSIVE PRICE
    239.00€
    106.96€ / 500mL
    Risparmia 132.04€
    55% Sconto
 Restricted Item
Spedizione stimata: 16-12-2026