Learn More
Brady™ ISO 7010 Sign - Fire hose reel
Product Code. 15694367 Shop All Brady Products
Change view
Click to view available options
Height (Metric):
148 mm
200 mm
315 mm
Width (Metric):
148 mm
200 mm
315 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Width (Metric) Unit Size
15694367 200 mm 200 mm Each
15684367 148 mm 148 mm Each
15614377 315 mm 315 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15694367 Supplier Brady™ Supplier No. 817047
  • ONLINE EXCLUSIVE PRICE
    10.50€
    9.42€ / pezzi 1
    Risparmia 1.08€
    10% Sconto
In stock
Prevent costly accidents with ISO 7010 safety signs for optimal impact and recognizability.
  • Polypropylene
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC F002-200x200-PP-CRD/1
Material Polypropylene
Quantity 1 Ea.
For Use With (Application) Indoor and Outdoor
Height (Metric) 200 mm
Thickness 1.501 mm
Width (Metric) 200 mm
Temperature Range (Metric) -40°C to 120°C
Shape Square
Languages Global
Color White on Red