Learn More
Brady™ ISO Safety Sign without Text - Warning: Low Temperature/Freezing Conditions
Product Code. 15871537 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W):
43 x 50 mm
87 x 100 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15871537 87 x 100 mm Each
15772652 43 x 50 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15871537 Supplier Brady™ Supplier No. 827799
  • ONLINE EXCLUSIVE PRICE
    9.40€
    8.45€ / pezzi 1
    Risparmia 0.95€
    10% Sconto
 Restricted Item
Spedizione stimata: 09-12-2026
Safety sign for hazard identification.
  • Sign compliant to ISO 7010
  • Improves workplace safety
Specifications
Alternate Part Number PIC W010-Tri 100-PE-SHEET/3
Finish Gloss
Material Laminated Polyester
Adhesive Type Permanent
Shape Triangular
Dimensions (L x W) 87 x 100 mm
Color Black on Yellow