Learn More
Isofield™ Isoclave CF Autoclavable Cleanroom Goggles - Grade A/B Compatible
Descrizione
- Close-fit autoclavable cleanroom goggles
- Anti-fog
- Indirect upper ventilation
- Non-vented base for added Cleanroom compliance
- Panoramic visibility
- Fits over standard prescription glasses
- Ergonomic frame with flexible nose skirt to ensure secure fit
- Premium anti-scratch
- Non-sterile - goggle must be processed prior to use
Specifica
Specifica
| Product Type | Cleanroom Goggles |
| Monouso | Single-use |
| Lens Material | Polycarbonate (Medical Grade) |
| Materiale del telaio | Silicone |