Learn More
Isofield™ Isoclave IV Autoclavable Cleanroom Goggles - Grade A/B Compatible
Descrizione
- Indirect upper and lower ventilation
- Panoramic visibility
- Fits over standard prescription glasses
- Ergonomic frame with flexible nose skirt
- Premium anti-scratch, optical grade, anti-fog lens
- Tested against EN 166 and ISO 16321-1
- Certified to PPE Cat II
- Non-sterile - Goggle must be processed prior to use
Specifica
Specifica
| Product Type | Cleanroom Goggles |
| Monouso | Single-use |
| Lens Material | Polycarbonate (Medical Grade) |
| Materiale del telaio | Thermoplastic Rubber (TPR) |