Learn More
Isopentyl Nitrite, TRC
Specifica
Specifica
| Materiale o nome chimico | Isopentyl Nitrite |
| CAS | 110-46-3 |
| Formula molecolare | C5 H11 N O2 |
| Quantità | 10 g |
| Sinonimo | Nitrous acid, 3-methylbutyl ester, Isopentyl alcohol, nitrite (8CI), 3-Methylbutanol nitrite, 3-Methylbutyl nitrite, Amyl nitrite, Aspiral, Isoamyl nitrite, Isopentyl nitrite, NSC 7903, Nitramyl, Nitrous acid, isopentyl ester, Pentyl nitrite, Vaporole |
| InChI Formula | InChI=1S/C5H11NO2/c1-5(2)3-4-8-6-7/h5H,3-4H2,1-2H3 |
| SMILES | CC(C)CCON=O |
| IUPAC Name | 3-methylbutyl nitrite |
| Molecular Weight (g/mol) | 117.15 |
| Peso formulazione | 117.079 |