Learn More
Isopropyl (2S)-7-Cyano-4-(pyridin-2-ylmethyl)-1,2,3,4-tetrahydrocyclopenta[b]indol-2-ylcarbamate, TRC
Product Code. 30138214
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138214 25 mg 25mg
30137962 50 mg 50mg
17904855 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30138214 Supplier Toronto Research Chemicals Supplier No. TRCI06570525MG
 Restricted Item
Spedizione stimata: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1029692-15-6
Molecular Formula C22 H22 N4 O2
Molecular Weight (g/mol) 374.44
Synonym 1-Methylethyl N-[(2S)-7-Cyano-1,2,3,4-tetrahydro-4-(2-pyridinylmethyl)cyclopent[b]indol-2-yl]carbamate, (S)-[7-Cyano-4-[(pyridin-2-yl)methyl]-1,2,3,4-tetrahydrocyclopenta[b]indol-2-yl]carbamic Acid Isopropyl Ester, LY 2452473, 1-Methylethyl Ester N-[(2S)-7-Cyano-1,2,3,4-tetrahydro-4-(2-pyridinylmethyl)cyclopent[b]indol-2-yl]carbamic Acid;
IUPAC Name isopropyl N-[(2S)-7-cyano-4-(2-pyridylmethyl)-2,3-dihydro-1H-cyclopenta[b]indol-2-yl]carbamate
SMILES CC(C)OC(=O)N[C@H]1Cc2c(C1)n(Cc3ccccn3)c4ccc(cc24)C#N
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C22H22N4O2/c1-14(2)28-22(27)25-17-10-19-18-9-15(12-23)6-7-20(18)26(21(19)11-17)13-16-5-3-4-8-24-16/h3-9,14,17H,10-11,13H2,1-2H3,(H,25,27)/t17-/m0/s1
Formula Weight 374.1743
Chemical Name or Material Isopropyl (2S)-7-Cyano-4-(pyridin-2-ylmethyl)-1,2,3,4-tetrahydrocyclopenta[b]indol-2-ylcarbamate