missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITPR3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA550688
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
The antibody detects endogenous levels of total ITPR3 protein.
ITPR3 Polyclonal antibody specifically detects ITPR3 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifica
ITPR3 | |
Polyclonal | |
Unconjugated | |
ITPR3 | |
inositol 1,4,5-triphosphate receptor 3; inositol 1,4,5-triphosphate receptor, type 3; inositol 1,4,5-trisphosphate receptor type 3; inositol 1,4,5-trisphosphate receptor, type 3; insP3R3; IP3 receptor; IP3 receptor isoform 3; IP3R; IP3R 3; Ip3r3; IP3R-3; IP3R3X; ITPR3; Itpr-3; Type 3 inositol 1,4,5-trisphosphate receptor; type 3 InsP3 receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3710 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q14573 | |
ITPR3 | |
Synthetic peptide corresponding to a region derived from internal residues of human inositol 1,4,5-trisphosphate receptor, type 3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |