Learn More
Avantor™ J.T. Baker™ BAKERBOND™ Florisil bulk packing material
Product Code. 15507654 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15507654 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15507654 Supplier Avantor™ Supplier No. 706100
  • ONLINE EXCLUSIVE PRICE
    108.63€
    72.80€ / 100g
    Risparmia 35.83€
    32% Sconto
 Restricted Item
Spedizione stimata: 29-12-2026
SPE sorbent for clean-up columns
  • Suitable for the determination of Hydrocarbon Oil Index according ISO 9377-2 and NEN 5733
Specifications
Adsorbent Type SPE
Description Florisil SPE sorbent for clean-up columns
Quantity 100 g