Learn More
Avantor™ J.T. Baker™ BAKERBOND Speedisk™ Expanded Extraction Station
Product Code. 15597694 Shop All J.T.Baker Products
Change view
Click to view available options
Pore Size:
185 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Unit Size
15597694 185 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15597694 Supplier Avantor™ Supplier No. 809506
  • ONLINE EXCLUSIVE PRICE
    12939.75€
    9533.64€ / pezzi 1
    Risparmia 3406.11€
    26% Sconto
 Restricted Item
Spedizione stimata: 21-05-2027
Can be used in reservoir, inverted, or remote sample feed modes
  • It includes a six-port vacuum manifold and the accessories needed to support the extraction of analyte by BAKERBOND Speedisk™ laminar extraction discs.
  • The manifold has a rectangular footprint and inter-port spacing to accommodate six, side-by-side, 1 L sample reservoirs
  • Each vacuum port has an individual open/close valve.
  • Includes extraction station, two remote sample adapters, two collection chambers and two vials, six reservoirs of 185 mL
Specifications
No. of Ports 6
Pore Size 185 mL
Size Expanded