Learn More
GPE Scientific™ J Young Gas Sample Tube with O-Ring Taps and Septum
Product Code. 15874764
Change view
Click to view available options
Measurement Range:
100 mL
125 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Unit Size
15874764 100 mL Each
15894764 125 mL Each
15824774 500 mL Each
15814774 250 mL Each
15884764 1000 mL Each
15804774 2000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15874764 Supplier GPE Scientific™ Supplier No. GSS/100
Spedizione stimata: 18-12-2026
GPE Scientific™ J Young Gas Sample Tube with O-Ring Taps and Septum is a gas collecting tube with O-ring valves on each end and a centrally located septum.
  • Cylindrical, oblong shape
  • Gas tight container
  • Gauged volume
  • Standard collecting tube with valved ends
  • Includes centralized septum
Specifications
Type Gas Sample Tube
Measurement Range 100 mL