missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ KleenGuard™ G80 Neoprene/Latex Chemical Resistant Gloves

Catalog Number 15336748
159.22 EUR valido fino al 2024-03-30
Usa il codice promo "21677" per avere il tuo prezzo promozionale.


Click to view available options
Size:
Large
Medium
Small
X-Large
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

G80 gloves have a natural rubber latex construction covered in neoprene to give a high resistance to acids, alkalis, oils and solvents. Ideal for removing paint, sealants and residues from a surface or degreasing machinery with a strong solvent.

  • Neoprene over natural rubber latex construction with cotton flock lining for comfort and perspiration absorption
  • High resistance to acids, oils, solvents and alkalis
  • Meets the EN 374-AKL standard and has a high level of dexterity, per EN420:2003 standard
  • Not suitable for those with Type 1 latex allergies
TRUSTED_SUSTAINABILITY
Certifications/Compliance (Europe) EN 420:2003, EN 374
Colore Blue, Yellow
Latex Free No
Materiale Neoprene, Latex
Size Large
Spessore (sistema imperiale) 27.5 mil
Spessore (sistema metrico) 0.69 mm
EN ISO 374-1 CHEMICAL LETTER CODES AKL
Stile polso guanto Straight
Grip Design Textured Fingertips
Materiale rivestimento interno Cotton Flock
Rivestimento interno Lined
Modello 38743
Serie Kleenguard™ G80
Lunghezza (sistema imperiale) 12 in.
Lunghezza (sistema metrico) 30.5 cm
Vedi altri risultati Mostra meno risultati
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'