Learn More
Velp Scientifica™ JLT Series Flocculators
Codice prodotto. 11791164 Sfoglia Tutto Velp Prodotti
Cambia vista
Click to view available options
Modello:
JLT4 - with 4 positions
JLT6 - with 6 positions
Wattage:
11 w
19 w
Peso (sistema metrico):
13 kg
17 kg
Larghezza (sistema metrico):
645 mm
935 mm
Dimensione della confezione:
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Modello Wattage Peso (sistema metrico) Larghezza (sistema metrico) unitSize
11791164 JLT4 - with 4 positions 11 w 13 kg 645 mm Pezzo
11701174 JLT6 - with 6 positions 19 w 17 kg 935 mm Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 11791164 Fornitore Velp Scientifica™ N. del fornitore F105A0108
  • ONLINE EXCLUSIVE PRICE
    1510.39€
    1395.00€ / pezzi 1
    Risparmia 115.39€
    7% Sconto
Visualizza tutte le promozioni
In stock
Aggiungi al carrello
Aggiungi al carrello
With multiple stirrers to allow standard conditions to be met during analysis, a basic requirement in order to obtain reliable and repeatable results. Velp Scientifica™ JLT Series Flocculators have digitally selectable speeds.
  • Microprocessor controlled operating time for unattended operation
  • Identical speed and time for all results for comparable results
  • Reliable and repeatable results from Velp stirring quality
  • Precise speed from an electronically controlled DC gear motor
  • High visibility LED displays for speed and time
  • Illuminated back panel to simplify sample observation
  • Stirring rods adjustable in height without tools
  • Strong resistance to chemical and mechanical corrosion
  • Available in 4- or 6-position models
Specifica
Speed 10 to 300 rpm
Tensione 100/240 V
Wattage 11 w
Altezza (sistema metrico) 347 mm
Larghezza (sistema metrico) 645 mm
Frequenza 50/60 Hz
Materiale Epoxy-painted Metal
Modello JLT4 - with 4 positions
Peso (sistema metrico) 13 kg