Descrizione
- Soft surface
- Easy to mould
- Comfortable to wear
- Low weight
Specifica
Specifica
| Certificazione/Conformità | EN 352-1 |
| Noise Reduction Rating (NRR) | 35 dB |
| Corded | No |
| Product Type | Ear Muffs |
| Monouso | Reusable |
| Metal Detectable | Yes |
| Headband Color | Black |
| Dielectric | No |
| Stile | Over-the-head |
| Foldable | Yes |