Learn More
Kaempferol, 98+%, Thermo Scientific Chemicals
Codice prodotto. 15433159
Click to view available options
Quantità:
10 mg
50 mg
Dimensione della confezione:
10mg
50mg
Codice prodotto. 15433159
Marca: Thermo Scientific Alfa Aesar J60373.MA
Richiesta di formato massivo o personalizzato
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Kaempferol, is used as an inhibitor of Fatty Acid Synthase, Cox-1 activity, and Topo I. It also Induces caspase-9-mediated apoptosis in a variety of cancer cell lines via downregulation of polo-like kinase 1 (PLK1) expression. Exhibits antioxidant activity and attenuates osteoclastic bone reabsorption in vitro. Also blocks EGF-induced histone H3Ser10 phosphorylation in mouse epidermal JB6 C141 cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Kaempferol, is used as an inhibitor of Fatty Acid Synthase, Cox-1 activity, and Topo I. It also Induces caspase-9-mediated apoptosis in a variety of cancer cell lines via downregulation of polo-like kinase 1 (PLK1) expression. Exhibits antioxidant activity and attenuates osteoclastic bone reabsorption in vitro. Also blocks EGF-induced histone H3Ser10 phosphorylation in mouse epidermal JB6 C141 cells.
Solubility
Soluble in DMSO or ethanol to 100mM
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Keep away fom strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 520-18-3
Formula molecolare C15H10O6
Molecular Weight (g/mol) 286.24
Numero MDL MFCD00016938
InChI Key IYRMWMYZSQPJKC-UHFFFAOYSA-N
Sinonimo kaempferol, kaempherol, kempferol, populnetin, rhamnolutein, robigenin, trifolitin, pelargidenolon, rhamnolutin, swartziol
PubChem CID 5280863
ChEBI CHEBI:28499
SMILES OC1=CC=C(C=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C=C2O1
Specifica
Materiale o nome chimico Kaempferol
Punto di fusione 276°C to 278°C
Colore Yellow
Quantità 10 mg
Beilstein 304401
Indice di Merck 14,5274
Informazioni di solubilità Soluble in DMSO or ethanol to 100mM
IUPAC Name 3,5,7-trihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one
Peso formulazione 286.24
Percent Purity ≥98%
Forma fisica Powder