Learn More
DiscoverX KILR™ RPMI 8226 Cell Pool
Codice prodotto. 17384859
Encompass
Cambia vista
Click to view available options
Quantità:
2 Vials
Dimensione della confezione:
2 fiale
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
17384859 2 Vials 2 fiale
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 17384859 Fornitore DiscoverX N. del fornitore 971026P039
The KILR™ RPMI 8226 Cell Pool allows for assay of effector-induced cell death by a variety of immune mechanisms. This cell pool ships as two vials of cryopreserved cells.
The KILR RPMI 8226 cell pool is a stable, engineered target cell pool which more closely reflects the heterogeneous native protein expression of tumor environments, used to measure direct cell death mediated by immunooncology treatments (e.g. checkpoint inhibitors, checkpoint agonists, bispecifics, CAR-T, and more) via Antibody Dependent Cell-mediated Cytotoxicity (ADCC), Complement Dependent Cytotoxicity (CDC), and Antibody Dependent Cell Phagocytosis (ADCP). This product uses EFC technology. The cell pool is a heterogeneous population of RPMI 8226 cells that overexpress a PL-tagged housekeeping gene. Once the cells have been lysed by effector cells, PL is released to the media, where EA contained in the detection kit produces an EFC signal proportional to the level of PL released. This allows for calculation of percent lysis of the KILR cell pool. This stable cell pool is provided as two vials of cryopreserved cells.
Specifica
Assay Entry Cytoxicity
Linea cellulare RPMI 8226
Da utilizzare con (applicazione) Cell Pools
Quantità 2 Vials