Learn More
DWK Life Sciences Kimble™ Black Phenolic Unlined Screw Thread Closures with Open Tops
Catalog Number p-73996
Click to view available options
Finitura collo:
15-425
24-400
Open top closures providing excellent chemical resistance
  • Phenolic
  • Open top
  • Autoclavable
  • Unlined
  • Black
Materiale chiusura Phenolic
Da utilizzare con (apparecchiatura) Bottle
Product Type Phenolic Unlined Screw Thread Closure
Colore Black
Materiale Phenolic Resin
N. per custodia 144 Cs.
Finitura collo 15-425
Closure Included Yes