Learn More
DWK Life Sciences Kimble™ Economy Columns Sample Diffusion Disc
Codice prodotto. 13476349
Click to view available options
Diametro (sistema metrico) interno:
1 cm
1.5 cm
2.5 cm
Dimensione della confezione:
Pezzo
Codice prodotto. 13476349
Marca: DWK Life Sciences 4204041000
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Flex-Column™ sample diffusion discs protect the gel bed from being disrupted during sample application whenever a flow adapter is not used.
  • A transparent acrylic ring is sealed to a 20um porosity HDPE disc which protects the top of the gel bed when the sample is manually applied
  • A crossbar on the top of the acrylic ring permits easy removal of the sample diffusion disc from the column.
TRUSTED_SUSTAINABILITY
Specifica
Tipo Sample Diffusion Disc
Diametro (sistema metrico) interno 1 cm
Da utilizzare con Kimble-Chase Economy Columns