Learn More
DWK Life Sciences KIMBLE™ KIMAX™ Pycnometer Bottle
Codice prodotto. 12737567 Sfoglia Tutto DWK Life Sciences Prodotti
Cambia vista
Click to view available options
Capacità (sistema metrico):
25 mL
50 mL
Altezza (sistema metrico):
175 mm
186 mm
Dimensione della confezione:
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Capacità (sistema metrico) Altezza (sistema metrico) Dimensione della confezione
12737567 50 mL 186 mm Pezzo
12727567 25 mL 175 mm Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 12737567 Fornitore DWK Life Sciences N. del fornitore 15123N50
  • ONLINE EXCLUSIVE PRICE
    288.00€
    242.35€ / pezzi 1
    Risparmia 45.65€
    15% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 05-03-2027
The KIMBLE™ KIMAX™ Pycnometer Bottle is designed for measuring and comparing the densities or specific gravities of liquids and solids. This non-mercury spirit-filled pycnometer includes a Standard Taper 10/18 thermometer ranging 14 to 38°C in 0.2° increments.
  • Designed for measuring and comparing the densities or specific gravities of liquids and solids
  • Non-mercury spirit-filled thermometer (15123N series); mercury-filled also available (15123R series)
  • Supplied with Standard Taper 10/18 thermometer with a range of 14 to 38°C in 0.2 degree increments
  • Thermometer is retested for accuracy within ±0.2 degrees C
  • Standard Taper 5/12 side tube cap has a small vent near the top
  • Body is conical for maximum stability
  • Capacity is within 10 percent of nominal shown on the flask
  • Each flask body is numbered for ready identification
Specifica
Tipo Pycnometer
Capacità (sistema metrico) 50 mL
Altezza (sistema metrico) 186 mm
Materiale Borosilicate Glass
Product Type Pycnometer Bottle