Learn More
DWK Life Sciences KIMBLE™ KONTES™ Adsorption/Ion-Exchange Columns
Product Code. 11897413
Change view
Click to view available options
Length (Metric):
300 mm
400 mm
500 mm
600 mm
Diameter (Metric) Inner:
11 mm
19 mm
22 mm
25 mm
41 mm
Column Capacity:
20 mL
110 mL
250 mL
790 mL
Includes:
Fritted disc
Without fritted disc
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Inner Column Capacity Includes Unit Size
11897413 400 mm 19 mm 110 mL Fritted disc Each
11887413 300 mm 11 mm 20 mL Without fritted disc Each
11807423 300 mm 22 mm 110 mL Fritted disc Each
11837423 600 mm 41 mm 790 mL Fritted disc Each
11827423 500 mm 25 mm 250 mL Fritted disc Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11897413 Supplier DWK Life Sciences Supplier No. 4203200224
  • ONLINE EXCLUSIVE PRICE
    101.00€
    83.18€ / pezzi 1
    Risparmia 17.82€
    17% Sconto
 Restricted Item
Spedizione stimata: 11-05-2027
KIMBLE™ KONTES™ Adsorption/Ion-Exchange Columns are available with or without a sealed-in fritted disc. Constructed from borosilicate glass conforming to USP Type I and ASTM E438 Type I Class A requirements for reliable chromatography separations.
  • Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • Suitable for adsorption and ion-exchange chromatography separations
  • K420320 Series columns feature a sealed-in coarse porosity (40 to 60 μm) fritted disc to support packing
  • K420300 Series columns use glass wool to support packing (packing not included)
Specifications
Pore Size 40 to 60 μm
Column Capacity 110 mL
Length (Metric) 400 mm
Material Borosilicate Glass
Includes Fritted disc
For Use With (Application) Many separations in adsorption and ion exchange chromatography
Type Adsorption/Ion Exchange Chromatography Column
Diameter (Metric) Inner 19 mm