Learn More
DWK Life Sciences KIMBLE™ KONTES™ Top Stopper for Incubation Flask
Product Code. 11853173 Shop All DWK Life Sciences Products
Change view
Click to view available options
No. per Pack:
144
Unit Size:
Pack of 144
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Pack Unit Size
11853173 144 Pack of 144
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11853173 Supplier DWK Life Sciences Supplier No. 8823100000
  • ONLINE EXCLUSIVE PRICE
    278.00€
    233.43€ / pezzi 144
    Risparmia 44.57€
    16% Sconto
 Restricted Item
Spedizione stimata: 10-02-2027
The KIMBLE™ KONTES™ Top Stopper for Incubation Flask fits both 10 and 25 mL sidearm incubation flasks, providing a double seal with an off-center hole for insertion of the center well. Sold separately in case quantities of 144.
  • Rubber top stopper designed for use with Kontes™ sidearm incubation flasks used in metabolic studies of biological materials
  • Provides a double seal and features an off-center hole for insertion of the center well
  • Compatible with both 10 mL and 25 mL sidearm incubation flasks
  • Enables reliable sealing while allowing syringe needle entry without disturbing established incubation conditions
  • Sidearm incubation flask system is suitable for measuring C14O2 yields, in vitro incubations, insulin bio-assays, persulfate oxidations, and distillation of volatile acids or bases
  • Top stoppers, sidearm stoppers, and center wells are available separately
  • Sold in case quantities of 144
Specifications
Material Rubber
For Use With (Application) For metabolic studies of biological materials
For Use With (Equipment) For incubation flask
No. per Pack 144
Product Type Stopper