Learn More
DWK Life Sciences Kimble™ Closures for 20mL Glass and Plastic Scintillation Vials
Durable screw thread closures
Marca: DWK Life Sciences 74525-22400
Dettagli aggiuntivi : Peso : 1.30000kg
Descrizione
- For 20mL Fisherbrand™ and Kimble scintillation vials, except 03-340-4B and 03-340-129. GPI 22-400 thread.
Specifica
Polyethylene | |
White | |
Urea |