Learn More
DWK Life Sciences KIMBLE™ Solid Borosilicate Glass Beads for Distillation Columns
Sfoglia Tutto DWK Life Sciences ProdottiDescrizione
- May be used as mixing beads, boiling stones, or packing for distillation columns
- Highly resistant to attack by most cold or hot liquids and/or vapors
- Solid borosilicate beads are durable and will not disintegrate or affect delicate compounds
- Packed in 1 lb. (approximately 0.45 kg) containers, 360 cm³ total volume
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Order Info
Packed in 1 lb. boxes
Specifica
Specifica
| Tipo | Beads |
| Materiale | Borosilicate Glass |
| Diametro (sistema metrico) | 4 mm |