Learn More
Thermo Scientific™ KingFisher™ Flex Deep Well heating block
Product Code. 12525977 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12525977 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12525977 Supplier Thermo Scientific™ Supplier No. 24075430
In stock
The KingFisher™ Flex 96 Deep-Well Heating Block is compatible with the KingFisher™ Flex Microtiter Deep-Well 96 Plate and the MagMAX™ Express-96 Deep-Well Plate when used on the KingFisher™ Flex Magnetic Particle Processor with 96 Deep-Well Head.
The KingFisher Flex 96 Deep-Well Heating Block is compatible with the KingFisher Flex Microtiter Deep-Well 96 Plate and the MagMAX Express-96 Deep-Well Plate when used on the KingFisher Flex Magnetic Particle Processor with 96 Deep-Well Head. Please note that the KingFisher Flex 96 Deep-Well Heating Block is not compatible with the KingFisher 96 microplate (200 μL).
TRUSTED_SUSTAINABILITY
Specifications
Format 96 Deep-well Plate
Capacity 96 Samples
For Use With (Equipment) KingFisher™ Flex
Product Line KingFisher
Quantity Each
Shipping Condition Room Temperature