Learn More
Thermo Scientific™ KingFisher™ Flex 96 heating block
Product Code. 12569437 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12569437 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12569437 Supplier Thermo Scientific™ Supplier No. 24075420
Spedizione stimata: 24-12-2026
The KingFisher™ Flex 96 KF Heating Block is compatible with the KingFisher™ 96 KF Microplate (200 μL) and the MagMAX™ Express-96 Standard Plate when used on the KingFisher™ Flex Magnetic Particle Processor with 96 Deep-Well Head.
The KingFisher™ Flex 96 KF Heating Block is compatible with the KingFisher™ 96 KF Microplate (200 μL) and the MagMAX™ Express-96 Standard Plate when used on the KingFisher™ Flex Magnetic Particle Processor with 96 Deep-Well Head. This heating block is also compatible with the KingFisher™ Flex Microtiter Deep Well 96 Plate and the MagMAX™ Express-96 Deep Well plates.
TRUSTED_SUSTAINABILITY
Specifications
Format 96-well Plate
Capacity 96 Samples
For Use With (Equipment) KingFisher™ Flex
Product Line KingFisher
Quantity Each
Shipping Condition Room Temperature