Learn More
Thermo Scientific™ KingFisher™ Systems Consumables
Product Code. 11520625 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
KingFisher plate 100 μL
KingFisher plate 200 μL
KingFisher tip comb
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
11520625 KingFisher tip comb Pack of 50
12673116 KingFisher plate 100 μL Pack of 50
12683116 KingFisher plate 200 μL Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11520625 Supplier Thermo Scientific™ Supplier No. 97002070
Black Friday Starts Here
Use promo code 27984
View all promotions
In stock
Optimize performance using Thermo Scientific™ KingFisher™Systems Consumables, ideal for magnetic particle processing.
Optimize performance using Thermo Scientific™ KingFisher™ Systems Consumables, specifically designed for use with the KingFisher System and ideal for magnetic particle processing because of their low binding affinity for biomolecules. The specially-designed, disposable tip combs and microplates, which are made of polypropylene, enable excellent recovery of magnetic beads.
  • Specifically designed for use with the KingFisher System
  • Ideal for magnetic particle processing because of their low binding affinity for biomolecules
  • The specially-designed, disposable tip combs and microplates are made of polypropylene
  • This enables excellent recovery of magnetic beads
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Magnetic particle processing
For Use With (Equipment) KingFisher System
Includes KingFisher tip comb
Quantity 50/Cs.