Learn More
DWK Life Sciences Kimble™ Kontes™ Disposaflex™ Polyethylene Disc
Product Code. 10749175
Change view
Click to view available options
Type:
Disc 30 to 50 μm
Disc, 20 μm
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10749175 Disc, 20 μm Pack of 100
11608228 Disc 30 to 50 μm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10749175 Supplier DWK Life Sciences Supplier No. 4201620020
  • ONLINE EXCLUSIVE PRICE
    76.75€
    60.53€ / pezzi 100
    Risparmia 16.22€
    21% Sconto
 Restricted Item
Spedizione stimata: 21-12-2026
For Disposaflex Chromatography Columns
  • Sold 100/pack.
Specifications
Type Disc, 20 μm
For Use With Disposaflex Chromatography Column
Material Polyethylene