Learn More
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (90mm)
Codice prodotto. 11867513
Cambia vista
Click to view available options
Volume (sistema metrico):
1 L
2 L
Dimensione della confezione:
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Volume (sistema metrico) unitSize
11867513 1 L Pezzo
11877513 2 L Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 11867513 Fornitore DWK Life Sciences N. del fornitore 9538250090
 Articolo soggetto a restrizioni
Spedizione stimata: 09-03-2027
Aggiungi al carrello
Aggiungi al carrello
Designed for large volume filtration analysis of corrosive liquids or samples with high particulate loads that would tend to clog a 47 mm diameter filter
This unit is also recommended for filtering large volumes of HPLC solvents.
  • The ground glass connection eliminates the possibility of phthalate contamination that can occur when using silicone or neoprene stoppers
      Each apparatus is supplied with 1000mL funnel; 90mm, 40-60 micron porosity, fritted glass support base; anodized aluminum clamp; and filtration flask
    • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifica
Materiale Glass
Tipo Microfiltration Assembly
Volume (sistema metrico) 1 L