missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KV3.1b (KCNC1) Polyclonal Antibody, Invitrogen™
Descrizione
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Voltage-gated K+ channels are important determits of neuronal membrane excitability. Moreover, differences in K+ channel expression patterns and densities contribute to the variations in action potential waveforms and repetitive firing patterns evident in different neuronal cell types (Maletic-Savatic et al., 1995; Pongs, 1999; Blaine and Ribera, 1998; Burger and Ribera, 1996). The Kv3.1 potassium channel is expressed at high levels in neurons that characteristically fire rapid trains of action potentials (Gan et al., 1999). Particularly high levels of this channel are found in neurons of the auditory brainstem. These neurons appear to participate in neural circuits that determine the intensity and timing of auditory stimuli and use this information to determine the location of sounds in space (von Hehn et al., 2004).
Specifica
Specifica
| Antigene | KV3.1b (KCNC1) |
| Applicazioni | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot |
| Classificazione | Polyclonal |
| Concentrazione | 0.8 mg/mL |
| Coniugato | Unconjugated |
| Formulazione | PBS with 1% BSA and 0.05% sodium azide; pH 7.4 |
| Gene | Kcnc1 |
| N. accesso geni | P15388, P25122 |
| Alias gene | Kcnc1 |
| Simboli geni | Kcnc1 |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?