Learn More
Abcam L-Amino Acid Assay Kit
Codice prodotto. 32173965 Sfoglia Tutto Abcam Prodotti
Cambia vista
Click to view available options
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto.
32173965 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 32173965 Fornitore Abcam N. del fornitore ab65347100Test
 Articolo soggetto a restrizioni
Spedizione stimata: 03-12-2026
Assay L-Amino Acid Assay in 30m in Biofluids, Cell/Tissue Extracts, Cell Culture Media with L-Amino Acid Assay Kit AB65347. Sensitivity ≤ 40 μM. Cited in 26 publications. Use with Microplate reader.
Assay L-Amino Acid Assay in 30m in Biofluids, Cell/Tissue Extracts, Cell Culture Media with L-Amino Acid Assay Kit AB65347. Sensitivity ≤ 40 μM. Cited in 26 publications. Use with Microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Quantità 100 Tests
Status giuridico RUO