missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteine hydrochloride monohydrate, MP Biomedicals™

Catalog Number 11403410
Click to view available options
Quantità:
1 kg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
7048-04-6
C3H10ClNO3S
175.63
MFCD00065606
QIJRTFXNRTXDIP-JIZZDEOASA-N
l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl
23462
(2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
C(C(C(=O)O)N)S.O.Cl
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
7048-04-6
C3H10ClNO3S
175.63
MFCD00065606
QIJRTFXNRTXDIP-JIZZDEOASA-N
l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl
23462
(2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
C(C(C(=O)O)N)S.O.Cl
Materiale o nome chimico L-Cysteine Hydrochloride Monohydrate
Nota nome Precursor in the food, pharmaceutical, personal care industries
Peso formulazione 175.64
Grado Cell Culture Reagent
Quantità 1 kg
Forma fisica Crystalline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'