missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine, Cell Culture Reagent, Thermo Scientific™

Catalog Number p-7042591
Click to view available options
Quantità:
10 g
100 g
500 g
1 kg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
71-00-1
C6H9N3O2
155.16
MFCD00064315
HNDVDQJCIGZPNO-UHFFFAOYNA-N
l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
6274
CHEBI:15971
NC(CC1=CN=CN1)C(O)=O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
71-00-1
C6H9N3O2
155.16
MFCD00064315
HNDVDQJCIGZPNO-UHFFFAOYNA-N
l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
6274
CHEBI:15971
NC(CC1=CN=CN1)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico L-Histidine
Punto di fusione ∽281°C (decomposition)
pH 7.0 to 8
Colore White
Beilstein 84088
Indice di Merck 14,4720
IUPAC Name (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
Peso formulazione 155.16
Grado Cell Culture Reagent
Quantità 10 g
Odore Odorless
Optical Rotation −39° (c=5 in Water)
Forma fisica Powder
Vedi altri risultati Mostra meno risultati
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'