Learn More
L-Homocysteine Thiolactone Hydrochloride, TRC
Codice prodotto. 30403517
Cambia vista
Click to view available options
Quantità:
5 g
Dimensione della confezione:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30403517 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30403517 Fornitore Toronto Research Chemicals N. del fornitore TRCH5919605G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico L-Homocysteine Thiolactone Hydrochloride
CAS 31828-68-9
Formula molecolare C4H8ClNOS
Quantità 5 g
Sinonimo (3S)-3-Aminodihydro-2(3H)-thiophenone Hydrochloride, (S)-3-Aminodihydro-2(3H)-thiophenone Hydrochloride, L-3-Aminodihydro-2(3H)-thiophenone Hydrochloride
InChI Formula InChI=1S/C4H7NOS.ClH/c5-3-1-2-7-4(3)6;/h3H,1-2,5H2;1H/t3-;/m0./s1
SMILES N[C@@H](CCS1)C1=O.Cl
Molecular Weight (g/mol) 153.63
Peso formulazione 153.0015