Learn More
L-Homocystine, MedChemExpress
Product Code. 16409356
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16409356 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16409356 Supplier Medchem Express Supplier No. HYW011690
 Restricted Item
Spedizione stimata: 01-12-2026
MedChemExpress L-Homocystine is the oxidized member of the L-homocysteine. Homocysteine is a pro-thrombotic factor, vasodilation impairing agent, pro-inflammatory factor and endoplasmatic reticulum-stress inducer used to study cardiovascular disease mechanisms.
L-Homocystine is the oxidized member of the L-homocysteine. Homocysteine is a pro-thrombotic factor, vasodilation impairing agent, pro-inflammatory factor and endoplasmatic reticulum-stress inducer used to study cardiovascular disease mechanisms.
Specifications
Chemical Name or Material L-Homocystine
CAS 626-72-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C8H16N2O4S2
Quantity 100 mg
Solubility Information H2O : 5 mg/mL (18.63 mM; ultrasonic and adjust pH to 3 with H2O) ∣DMSO : 1 mg/mL (3.73 mM; ultrasonic and adjust pH to 5 with HCl)
SMILES OC([C@@H](N)CCSSCC[C@H](N)C(O)=O)=O
Molecular Weight (g/mol) 268.35
Formula Weight 268.35
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only