missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Prolinamide, 98%, Thermo Scientific Chemicals

Catalog Number p-7048168
44.46 EUR valido fino al 2024-03-29
Usa il codice promo "21615" per avere il tuo prezzo promozionale.


Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7531-52-4
C5H11N2O
115.16
MFCD00005253
VLJNHYLEOZPXFW-BYPYZUCNSA-O
l-prolinamide, prolinamide, h-pro-nh2, s-pyrrolidine-2-carboxamide, s-prolinamide, 2s-pyrrolidine-2-carboxamide, l-prolineamide, l-proline amide, l---prolinamide, s-2-pyrrolidinecarboxamide
111306
CHEBI:21374
NC(=O)[C@@H]1CCC[NH2+]1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7531-52-4
C5H11N2O
115.16
MFCD00005253
VLJNHYLEOZPXFW-BYPYZUCNSA-O
l-prolinamide, prolinamide, h-pro-nh2, s-pyrrolidine-2-carboxamide, s-prolinamide, 2s-pyrrolidine-2-carboxamide, l-prolineamide, l-proline amide, l---prolinamide, s-2-pyrrolidinecarboxamide
111306
CHEBI:21374
NC(=O)[C@@H]1CCC[NH2+]1

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. Also it is a efficient organocatalysts for aldol reactions on water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. Also it is a efficient organocatalysts for aldol reactions on water.

Solubility
Soluble in water and ethanol (50 mg/ml).

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico L-(-)-Prolinamide
Punto di fusione 95°C to 104°C
Quantità 1 g
Beilstein 80807
Informazioni di solubilità Soluble in water and ethanol (50mg/ml).
Optical Rotation −105° (c=2 in Ethanol)
IUPAC Name (2S)-pyrrolidine-2-carboxamide
Peso formulazione 114.15
Percent Purity 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'