Learn More
L-Valine 2-Hydroxyethyl Ester Tosylate, TRC
Product Code. 30450344
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30450344 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30450344 Supplier Toronto Research Chemicals Supplier No. TRCV09432025MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-hydroxyethyl valinate para-toluenesulfonate salt
CAS 86150-61-0
Molecular Formula C7 H15 N O3 . C7 H8 O3 S
Quantity 25 mg
Synonym L-Valine','2-hydroxyethyl ester','4-methylbenzenesulfonate (salt) (9CI), 2-Hydroxyethyl L-Valinate para-Toluenesulfonate
InChI Formula InChI=1S/C7H15NO3.C7H8O3S/c1-5(2)6(8)7(10)11-4-3-9;1-6-2-4-7(5-3-6)11(8,9)10/h5-6,9H,3-4,8H2,1-2H3;2-5H,1H3,(H,8,9,10)/t6-;/m0./s1
SMILES CC(C)[C@H](N)C(=O)OCCO.Cc1ccc(cc1)S(=O)(=O)O
IUPAC Name 2-hydroxyethyl (2S)-2-amino-3-methylbutanoate;4-methylbenzenesulfonic acid
Molecular Weight (g/mol) 333.4
Formula Weight 333.1246