Learn More
L-xylo-2-Hexulosonic Acid Methyl Ester, TRC
Specifica
Specifica
| Chemical Name or Material | Methyl L-Sorbosonate |
| CAS | 3031-98-9 |
| Molecular Formula | C7 H12 O7 |
| Quantity | 100 mg |
| Synonym | L-xylo-2-Hexulosonic acid','methyl ester, Gulonic acid','2-keto-','methyl ester','L- (6CI), L-xylo-Hexulosonic acid','methyl ester (7CI,8CI), 2-keto-L-Gulonic acid methyl ester, Methyl 2-keto-L-gulonate, Methyl L-xylo-hexulosonate, NSC 163924, Methyl L-Sorbosonate, Methyl L-xylo-Hex-2-ulosonate, Ascorbic Acid Imp. D (EP) |
| InChI Formula | InChI=1S/C7H12O7/c1-14-7(13)6(12)5(11)4(10)3(9)2-8/h3-5,8-11H,2H2,1H3/t3-,4+,5-/m0/s1 |
| SMILES | COC(=O)C(=O)[C@@H](O)[C@H](O)[C@@H](O)CO |
| IUPAC Name | methyl (3S,4R,5S)-3,4,5,6-tetrahydroxy-2-oxohexanoate |
| Molecular Weight (g/mol) | 208.166 |
| Formula Weight | 208.0583 |