Descrizione
For use with Labconco Protector XVS Ventilation Stations
Ductwork
- Standard 6 in. dia. (15.2cm) Type I PVC duct can be cut without special tools; secured with solvent cement (Female duct coupling required to join two sections. See Cat. No. 16-304-12)
- 90° elbow (Cat. No. 16-304-11) has belled end connections to receive PVC duct directly
- Flexible duct connection reduces vibration between blower and PVC ductwork; supplied with two installation clamps
- Powder-coated steel blower transition adapter allows round PVC duct to be connected to the exhaust side of the blower
- PVC zero-pressure weathercap (Cat. No. 16-304-14) connects directly to the duct for vertical discharge of effluent air and dispersion away from the building
- Cap keeps out rain and snow but adds little static pressure to the exhaust system
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | Protector XVS Ventilation Station |
| Peso (sistema imperiale) | 25 lb. |
| Peso (sistema metrico) | 11.3 Kg |
| Product Type | PVC Duct |
| Materiale | PVC |