Learn More
Labconco™ Protector XVS™ Zero–Pressure PVC Weathercap for Ventilation Stations
Sfoglia Tutto Labconco Corporation ProdottiDescription
- For use with Labconco Protector XVS Ventilation Stations
- PVC zero-pressure weathercap connects directly to duct for vertical discharge of effluent air and dispersion away from building
- Cap keeps out rain and snow but adds little static pressure to exhaust system
Specifications
Specifications
| Da utilizzare con (apparecchiatura) | Protector VS Ventilation Station |
| Altezza (sistema imperiale) | 36 in. |
| Peso (sistema imperiale) | 20 lb. |
| Peso (sistema metrico) | 9.1 Kg |
| Include | Weathercap |
| Dimensions | 6 in. dia. round PVC duct |
| Materiale | PVC |
| Da utilizzare con (applicazione) | Ducting |