Learn More
Labconco™ SpillStopper™ Work Surface for 4 Ft. Protector™ Premier™ Hood
Codice prodotto. 17621037 Sfoglia Tutto Labconco Corporation Prodotti
Cambia vista
Click to view available options
Stile di taglio:
No Cutouts
Right Rear
Dimensione della confezione:
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Stile di taglio Dimensione della confezione
17621037 Right Rear Pezzo
16338705 No Cutouts Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 17621037 Fornitore Labconco™ N. del fornitore 9500401
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Labconco SpillStopper™ Work Surface is dished and contoured to conform to the interior liner to contain spills.
  • Corrosion-resistant epoxy resins molded
  • Front edge with large radius to aerodynamically direct airflow into hood
  • Pre-drilled holes for venting and plumbing pass-throughs provided
  • Some surfaces with cutouts for surface-mounted service fixture and/or 6 x 3 in. (15.2 x 7.6cm) cup sinkcup sink
  • Flat work surfaces may also be coupled with adjustable height base stand to provide auxiliary workspace
Order Info
Service fixture and cup sink sold separately
Specifica
Da utilizzare con (apparecchiatura) Protector XL Laboratory Hood
Include Right Rear
Product Type SpillStopper Work Surface (4 ft.)
Materiale Epoxy Resin
Da utilizzare con (applicazione) Laboratory Fume Hood Work
Stile di taglio Right Rear