Learn More
Labconco™ Trap Insert Canister with Stand for Precise™ Glove Boxes
Product Code. 11751479 Shop All Labconco Corporation Products
Change view
Click to view available options
Material:
Polypropylene
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
11751479 Polypropylene Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11751479 Supplier Labconco™ Supplier No. 7815301
  • ONLINE EXCLUSIVE PRICE
    550.00€
    523.96€ / pezzi 1
    Risparmia 26.04€
    4% Sconto
 Restricted Item
Spedizione stimata: 17-12-2026
Clear polypropylene canister
  • Accommodates any of the following Chemical Trap Inserts: Acid Trap (Mfr. No. 7814800), Moisture Trap (Mfr. No. 7814900), Radioisotope Trap (Mfr. No. 7815000), Solvent Trap (Mfr. No. 7815200), or Ammonia Trap (Mfr. No. 79956000), each sold separately
  • For use with Precise™ Controlled Atmosphere and Basic Glove Boxes,as well as Protector™ Combination and Controlled Atmosphere Glove Boxes
Specifications
Color Clear
Case Material Polypropylene
Material Polypropylene
Maximum Pressure 1 bar
For Use With (Equipment) 7814800 Acid trap, 7814900 moisture trap, 7815000 Radioisotope trap, 7815200 Solvent trap or 7995600 Ammonia trap insert
Includes Stand
Type Canister
Weight (English) 5 lb.
Weight (Metric) 2.3 kg