Learn More
Labconco™ Tube Holders for Fast-Freeze™ and Lyph-Lock™ Flasks
Product Code. 10316372 Shop All Labconco Corporation Products
Change view
Click to view available options
Height (Metric):
13.9 cm
14.4 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
10316372 13.9 cm Each
10123292 14.4 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10316372 Supplier Labconco™ Supplier No. 7442700
  • ONLINE EXCLUSIVE PRICE
    348.00€
    282.31€ / pezzi 1
    Risparmia 65.69€
    18% Sconto
View all promotions
 Restricted Item
Spedizione stimata: 24-12-2026
Holds tubes in upright position for freeze drying.
Hold tubes in upright position inside 900mL or larger Fast-Freeze Flasks and 750mL or larger Lyph-Lock Flasks Stainless steel
Specifications
Type Tube Holder
Capacity (Metric) 750, 900 mL
Material Stainless Steel
For Use With (Application) Hold tubes in upright position
For Use With (Equipment) 900ml or larger fast freeze™ flasks and 750ml or larger lyph lock flasks
Diameter (English) 3.5 in.
Diameter (Metric) 8.33 cm
Height (English) 5 in.
Height (Metric) 13.9 cm
Weight (English) 0.5 lb.
Weight (Metric) 0.5 kg