Learn More
Labconco™ VACUUBRAND™ Hybrid™ Vacuum Pump Oil
Product Code. 16205181 Shop All Labconco Corporation Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16205181 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16205181 Supplier Labconco™ Supplier No. 7584300
  • ONLINE EXCLUSIVE PRICE
    181.00€
    171.79€ / 1L
    Risparmia 9.21€
    5% Sconto
 Restricted Item
Spedizione stimata: 05-01-2027
For VACUUBRAND pumps
VACUUBRAND™ B-Oil. One. liter bottle. Replacement oil for VACUUBRAND HYBRID™ Vacuum Pumps 7584000 and 7584002.
Specifications
Product Type Oil
Format Oil
For Use With (Equipment) Hybrid Vacuum Pumps
Quantity 1 L
Volume (English) 33.8 oz.
Volume (Metric) 1 L