Learn More
SHP Steriltechnik AG Laboklav 25 Wire Tray
Product Code. 31908801
Change view
Click to view available options
For Use With (Equipment):
Laboklav 25
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
31908801 Laboklav 25 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31908801 Supplier SHP Steriltechnik AG Supplier No. 30.4220.001.025
Spedizione stimata: 22-01-2027
Stainless steel wire tray provides stable placement and efficient organisation of instruments while ensuring optimal steam circulation during sterilisation.
  • For Laboklav 25
  • Stainless steel
  • WxHxD (mm): 178 x 30 x 425
Specifications
Capacity (Metric) 2 L
For Use With (Equipment) Laboklav 25
Model Tray, Laboklav 25
Product Type Tray
Sterility Non-sterile