Learn More
Lenz Laborglasintrumente Laboratory Bottles
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Marca:  Lenz Laborglasintrumente 07105094
Codice prodotto. 10020421
  • ONLINE EXCLUSIVE PRICE
    973.00€
    725.84€ / pezzi 1
    Risparmia 247.16€
    25% Sconto
Spedizione stimata: 27-10-2026
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Acc. to ISO 4796, with DIN thread
  • Material: DURAN™
  • With easy-to-read scale and large labelling field for easy marking
  • With screw cap and pouring ring (material: PP, blue; temperature resistance: +140 °C)