Learn More
Bochem™ Laboratory Pot
Codice prodotto. 10539242
Cambia vista
Click to view available options
Capacity: 20L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto.
10539242 Capacity: 20L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 10539242 Fornitore Bochem™ N. del fornitore 8367
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Bochem™ Laboratory Pot are constructed of Stainless Steel for durability.
  • Stainless Steel
Specifica
Diametro (sistema metrico) interno 320mm
Altezza (sistema metrico) 270mm
Capacità (sistema metrico) 20 L
Materiale Stainless Steel (18/10)